siRNA Details
| Gene Symbol: NAIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004536.2 | NP_004527.2 | 4 | 858 | ||
| GenBank mRNA | AK309627.1 | - | 2 | 215 | ||
| BC136273.1 | - | 2 | 195 | 9020260 | ||
| BC143761.1 | - | 2 | 195 | 9052275 | ||
| U19251.1 | - | 4 | 431 | |||

NAIP
Genomic Map