siRNA Details
| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022589.1 | NP_072111.1 | 2 | 150 | ||
| GenBank mRNA | AJ271442.1 | - | 2 | 150 | ||
| BC085733.1 | - | 2 | 170 | 7122828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan2| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022589.1 | NP_072111.1 | 2 | 150 | ||
| GenBank mRNA | AJ271442.1 | - | 2 | 150 | ||
| BC085733.1 | - | 2 | 170 | 7122828 | ||