siRNA Details
| Gene Symbol: Pnpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022601.2 | NP_072123.1 | 1, 2 | 278 | ||
| GenBank mRNA | BC087016.1 | - | 1, 2 | 278 | 7128621 | |
| U91561.1 | - | 1, 2 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnpo| Gene Symbol: Pnpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022601.2 | NP_072123.1 | 1, 2 | 278 | ||
| GenBank mRNA | BC087016.1 | - | 1, 2 | 278 | 7128621 | |
| U91561.1 | - | 1, 2 | 269 | |||