siRNA Details
| Gene Symbol: Ilkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022606.1 | NP_072128.1 | 3, 4 | 231 | ||
| GenBank mRNA | AF095927.1 | - | 3, 4 | 231 | ||
| BC062010.1 | - | 3, 4 | 233 | 5621497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ilkap| Gene Symbol: Ilkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022606.1 | NP_072128.1 | 3, 4 | 231 | ||
| GenBank mRNA | AF095927.1 | - | 3, 4 | 231 | ||
| BC062010.1 | - | 3, 4 | 233 | 5621497 | ||