siRNA Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022615.1 | NP_072137.1 | 3 | 116 | ||
| GenBank mRNA | AF140782.1 | - | 3 | 116 | ||
| AY325188.1 | - | 2 | 105 | |||
| Gene Symbol: LOC499179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759598.1 | XP_008757820.1 | 3 | 254 | ||

Top1
Genomic Map