siRNA Details
| Gene Symbol: Slc5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022590.2 | NP_072112.2 | 7 | 739 | ||
| GenBank mRNA | AB070849.1 | - | 6 | 874 | ||
| AB070850.1 | - | 6 | 874 | |||
| AB070976.1 | - | 7 | 734 | |||
| U29881.1 | - | 7 | 739 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a2| Gene Symbol: Slc5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022590.2 | NP_072112.2 | 7 | 739 | ||
| GenBank mRNA | AB070849.1 | - | 6 | 874 | ||
| AB070850.1 | - | 6 | 874 | |||
| AB070976.1 | - | 7 | 734 | |||
| U29881.1 | - | 7 | 739 | |||