siRNA Details
| Gene Symbol: Tep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022591.2 | NP_072113.1 | 2 | 698 | ||
| GenBank mRNA | U89282.1 | - | 2 | 699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tep1| Gene Symbol: Tep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022591.2 | NP_072113.1 | 2 | 698 | ||
| GenBank mRNA | U89282.1 | - | 2 | 699 | ||