siRNA Details
| Gene Symbol: EPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000537.1 | XP_016856026.1 | 1, 2 | 261 | ||
| GenBank mRNA | AK296788.1 | - | 1, 2 | 226 | ||
| BC037166.2 | - | 1, 2 | 207 | 5090654 | ||
| CB991812.1 | - | 1, 2 | 271 | |||
| EU826606.1 | - | 1, 2 | 89 | |||
| HY005615.1 | - | 1, 2 | 267 | |||
| M59371.1 | - | 1, 2 | 202 | |||

EPHA2
Genomic Map