siRNA Details
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022597.2 | NP_072119.2 | 7, 8 | 800 | ||
| GenBank mRNA | BC072490.1 | - | 7, 8 | 763 | 7097817 | |
| FQ212977.1 | - | 3, 4 | 311 | |||
| M11305.1 | - | 4, 5 | 464 | |||
| X82396.1 | - | 7, 8 | 729 | |||

Ctsb
Genomic Map