siRNA Details
| Gene Symbol: Fgfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022603.1 | NP_072125.1 | 1 | 202 | ||
| GenBank mRNA | AF142758.1 | - | 1 | 202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgfbp1| Gene Symbol: Fgfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022603.1 | NP_072125.1 | 1 | 202 | ||
| GenBank mRNA | AF142758.1 | - | 1 | 202 | ||