siRNA Details
| Gene Symbol: Slc7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134686.2 | NP_001128158.1 | 4 | 824 | ||
| NM_022619.3 | NP_072141.2 | 4 | 824 | |||
| XM_008771281.1 | XP_008769503.1 | 4 | 5903 | |||
| XM_008771282.1 | XP_008769504.1 | 5 | 783 | |||
| XM_008771283.1 | XP_008769505.1 | 4 | 5903 | |||
| GenBank mRNA | AF245001.1 | - | 4 | 825 | ||
| AF245002.1 | - | 4 | 825 | |||
| U53927.1 | - | 1 | 47 | |||

Slc7a2
Genomic Map