siRNA Details
| Gene Symbol: Tpc1808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022625.2 | NP_072147.1 | 960 | |||
| GenBank mRNA | AF078811.2 | - | 960 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpc1808| Gene Symbol: Tpc1808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022625.2 | NP_072147.1 | 960 | |||
| GenBank mRNA | AF078811.2 | - | 960 | |||