siRNA Details
| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022513.2 | NP_071958.1 | 4 | 497 | ||
| XM_008770056.1 | XP_008768278.1 | 4 | 371 | |||
| GenBank mRNA | D89375.1 | - | 4 | 448 | ||
| FQ209476.1 | - | 4 | 465 | |||
| FQ209486.1 | - | 4 | 531 | |||
| FQ209579.1 | - | 4 | 552 | |||
| FQ210801.1 | - | 4 | 539 | |||
| FQ212608.1 | - | 4 | 508 | |||
| FQ218295.1 | - | 4 | 510 | |||
| U38419.1 | - | 4 | 500 | |||

Sult1b1
Genomic Map