siRNA Details
| Gene Symbol: Des | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022531.1 | NP_071976.1 | 2 | 671 | ||
| GenBank mRNA | BC061872.1 | - | 2 | 664 | 5598951 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Des| Gene Symbol: Des | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022531.1 | NP_071976.1 | 2 | 671 | ||
| GenBank mRNA | BC061872.1 | - | 2 | 664 | 5598951 | |