siRNA Details
| Gene Symbol: Metap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022539.1 | NP_071984.1 | 10 | 1516 | ||
| GenBank mRNA | BC070892.1 | - | 10 | 1432 | 7099147 | |
| BC091150.1 | - | 10 | 1531 | 7122560 | ||
| L10652.1 | - | 10 | 1516 | |||
| Gene Symbol: RGD1560341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574154.6 | XP_574154.2 | 1 | 1547 | ||

Metap2
Genomic Map