siRNA Details
| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022548.1 | NP_071993.1 | 5 | 826 | ||
| GenBank mRNA | Y13148.1 | - | 5 | 826 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmat3| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022548.1 | NP_071993.1 | 5 | 826 | ||
| GenBank mRNA | Y13148.1 | - | 5 | 826 | ||