siRNA Details
| Gene Symbol: Tkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022592.1 | NP_072114.1 | 5 | 534 | ||
| XM_006252661.1 | XP_006252723.1 | 4 | 566 | |||
| GenBank mRNA | U09256.1 | - | 5 | 534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tkt| Gene Symbol: Tkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022592.1 | NP_072114.1 | 5 | 534 | ||
| XM_006252661.1 | XP_006252723.1 | 4 | 566 | |||
| GenBank mRNA | U09256.1 | - | 5 | 534 | ||