siRNA Details
| Gene Symbol: Adcy5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022600.1 | NP_072122.1 | 20, 21 | 4101 | ||
| XM_008768744.1 | XP_008766966.1 | 20 | 5306 | |||
| GenBank mRNA | M96159.1 | - | 20, 21 | 4101 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adcy5| Gene Symbol: Adcy5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022600.1 | NP_072122.1 | 20, 21 | 4101 | ||
| XM_008768744.1 | XP_008766966.1 | 20 | 5306 | |||
| GenBank mRNA | M96159.1 | - | 20, 21 | 4101 | ||