siRNA Details
| Gene Symbol: Fgfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022603.1 | NP_072125.1 | 1 | 296 | ||
| XM_006251078.2 | XP_006251140.1 | 2 | 430 | |||
| XM_006251079.2 | XP_006251141.1 | 2 | 352 | |||
| GenBank mRNA | AF142758.1 | - | 1 | 296 | ||

Fgfbp1
Genomic Map