siRNA Details
| Gene Symbol: Hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022605.1 | NP_072127.1 | 10 | 1303 | ||
| GenBank mRNA | AF184967.1 | - | 10 | 1303 | ||
| AF359508.1 | - | 10 | 1330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpse| Gene Symbol: Hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022605.1 | NP_072127.1 | 10 | 1303 | ||
| GenBank mRNA | AF184967.1 | - | 10 | 1303 | ||
| AF359508.1 | - | 10 | 1330 | |||