siRNA Details
| Gene Symbol: Phka1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022626.1 | NP_072148.1 | 24, 25 | 2699 | ||
| XM_006257101.2 | XP_006257163.1 | 24, 25 | 3026 | |||
| XM_006257102.2 | XP_006257164.1 | 23, 24 | 2849 | |||
| GenBank mRNA | AF197561.1 | - | 24, 25 | 2699 | ||

Phka1
Genomic Map