siRNA Details
| Gene Symbol: Grm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022666.1 | NP_073157.1 | 2005 | |||
| GenBank mRNA | M90518.1 | - | 2005 | |||
| M92077.1 | - | 1220 | ||||
| U47331.1 | - | 2005 | ||||
More Information
Other Species Targeted
Set Membership: |
|

Grm4
Genomic Map