siRNA Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022667.2 | NP_073158.2 | 10, 11 | 1549 | ||
| XM_006243655.1 | XP_006243717.1 | 11, 12 | 1531 | |||
| GenBank mRNA | M64862.1 | - | 10, 11 | 1550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco2a1| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022667.2 | NP_073158.2 | 10, 11 | 1549 | ||
| XM_006243655.1 | XP_006243717.1 | 11, 12 | 1531 | |||
| GenBank mRNA | M64862.1 | - | 10, 11 | 1550 | ||