siRNA Details
| Gene Symbol: Scg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022669.1 | NP_073160.1 | 1 | 806 | ||
| XM_006245148.2 | XP_006245210.1 | 2 | 996 | |||
| XM_008767195.1 | XP_008765417.1 | 2 | 1130 | |||
| GenBank mRNA | AY168445.1 | - | 1 | 464 | ||
| BC061549.1 | - | 2 | 848 | 6919444 | ||
| M93669.1 | - | 1 | 806 | |||
| X13618.1 | - | 1 | 806 | |||

Scg2
Genomic Map