siRNA Details
| Gene Symbol: Ntsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022695.2 | NP_073186.1 | 1, 2 | 658 | ||
| XM_006239918.2 | XP_006239980.1 | 1, 2 | 626 | |||
| GenBank mRNA | AY946024.1 | - | 1, 2 | 620 | ||
| X97121.1 | - | 1, 2 | 660 | |||
More Information
Set Membership: |
|

Ntsr2
Genomic Map