siRNA Details
| Gene Symbol: Unc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022862.1 | NP_074053.1 | 2 | 576 | ||
| GenBank mRNA | U24071.1 | - | 2 | 576 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc13b| Gene Symbol: Unc13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022862.1 | NP_074053.1 | 2 | 576 | ||
| GenBank mRNA | U24071.1 | - | 2 | 576 | ||