siRNA Details
| Gene Symbol: Pcdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022868.1 | NP_074059.1 | 1 | 161 | ||
| XM_006252375.2 | XP_006252437.1 | 1 | 827 | |||
| XM_008770897.1 | XP_008769119.1 | 2 | 590 | |||
| GenBank mRNA | AB026154.1 | - | 1 | 161 | ||
| BC128737.1 | - | 1 | 350 | |||

Pcdh8
Genomic Map