siRNA Details
| Gene Symbol: Dnaja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022934.1 | NP_075223.1 | 2 | 201 | ||
| XM_006238099.1 | XP_006238161.1 | 2 | 299 | |||
| GenBank mRNA | BC062009.1 | - | 2 | 158 | 5621557 | |
| FQ216722.1 | - | 2 | 190 | |||
| U53922.1 | - | 2 | 201 | |||

Dnaja1
Genomic Map