siRNA Details
| Gene Symbol: Stx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022939.2 | NP_075228.2 | 2 | 248 | ||
| XM_006239066.2 | XP_006239128.1 | 2 | 292 | |||
| GenBank mRNA | AF035632.1 | - | 2 | 164 | ||
| CB581551.1 | - | 2 | 237 | |||
| CK602227.1 | - | 2 | 248 | |||

Stx12
Genomic Map