siRNA Details
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767428.1 | XP_008765650.1 | 1 | 837 | ||
GenBank mRNA | U67137.1 | - | 3 | 857 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767428.1 | XP_008765650.1 | 1 | 837 | ||
GenBank mRNA | U67137.1 | - | 3 | 857 |