siRNA Details
Gene Symbol: Bid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022684.1 | NP_073175.1 | 4 | 433 | ||
GenBank mRNA | AF136282.1 | - | 2 | 291 | ||
AF259503.1 | - | 4 | 433 | |||
D89804.1 | - | 389 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022684.1 | NP_073175.1 | 4 | 433 | ||
GenBank mRNA | AF136282.1 | - | 2 | 291 | ||
AF259503.1 | - | 4 | 433 | |||
D89804.1 | - | 389 |