siRNA Details
| Gene Symbol: Rab5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022692.1 | NP_073183.1 | 324 | |||
| GenBank mRNA | AF072935.1 | - | 324 | |||
| BC161848.1 | - | 650 | 9025210 | |||
| Gene Symbol: Rab5al1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751377.3 | XP_003751425.1 | 1 | 556 | ||

Rab5a
Genomic Map