siRNA Details
| Gene Symbol: Crhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022714.1 | NP_073205.1 | 3 | 498 | ||
| GenBank mRNA | EF078964.1 | - | 3 | 283 | ||
| EF078965.1 | - | 3 | 283 | |||
| U16253.1 | - | 3 | 498 | |||
More Information
Set Membership: |
|

Crhr2
Genomic Map