siRNA Details
| Gene Symbol: TRIP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004237.3 | NP_004228.1 | 13 | 1798 | ||
| GenBank mRNA | BC000404.2 | - | 13 | 1763 | 2822981 | |
| BC019294.1 | - | 13 | 1763 | 2822981 | ||
| U96131.1 | - | 13 | 1698 | |||

TRIP13
Genomic Map