siRNA Details
| Gene Symbol: Crisp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022859.1 | NP_074050.1 | 5, 6 | 393 | ||
| GenBank mRNA | M31173.1 | - | 5, 6 | 393 | ||
| X04643.1 | - | 5, 6 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crisp1| Gene Symbol: Crisp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022859.1 | NP_074050.1 | 5, 6 | 393 | ||
| GenBank mRNA | M31173.1 | - | 5, 6 | 393 | ||
| X04643.1 | - | 5, 6 | 365 | |||