siRNA Details
| Gene Symbol: Map1lc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022867.2 | NP_074058.2 | 4 | 1328 | ||
| GenBank mRNA | AY206670.1 | - | 1 | 390 | ||
| AY392036.1 | - | 4 | 1328 | |||
| BC083556.1 | - | 4 | 1308 | 7189855 | ||
| U05784.1 | - | 5 | 443 | |||

Map1lc3b
Genomic Map