siRNA Details
| Gene Symbol: Nolc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022869.1 | NP_074060.1 | 3 | 291 | ||
| XM_006231560.2 | XP_006231622.1 | 3 | 342 | |||
| GenBank mRNA | BC126088.1 | - | 3 | 256 | ||
| M94287.1 | - | 3 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nolc1| Gene Symbol: Nolc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022869.1 | NP_074060.1 | 3 | 291 | ||
| XM_006231560.2 | XP_006231622.1 | 3 | 342 | |||
| GenBank mRNA | BC126088.1 | - | 3 | 256 | ||
| M94287.1 | - | 3 | 291 | |||