siRNA Details
| Gene Symbol: Clpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022947.1 | NP_075236.1 | 1, 2 | 455 | ||
| GenBank mRNA | AB027570.1 | - | 1, 2 | 455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clpb| Gene Symbol: Clpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022947.1 | NP_075236.1 | 1, 2 | 455 | ||
| GenBank mRNA | AB027570.1 | - | 1, 2 | 455 | ||