siRNA Details
Gene Symbol: H2afz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022674.1 | NP_073165.1 | 5 | 419 | ||
GenBank mRNA | BC060564.1 | - | 5 | 484 | 6922321 | |
BC086348.1 | - | 5 | 464 | 7302603 | ||
FQ213837.1 | - | 5 | 478 | |||
FQ220829.1 | - | 5 | 480 | |||
FQ221791.1 | - | 5 | 478 | |||
FQ222470.1 | - | 5 | 478 | |||
FQ224994.1 | - | 5 | 481 | |||
FQ227672.1 | - | 5 | 480 | |||
FQ229641.1 | - | 5 | 480 | |||
FQ229746.1 | - | 5 | 480 | |||
FQ232362.1 | - | 5 | 480 | |||
FQ233167.1 | - | 5 | 480 | |||
FQ234498.1 | - | 5 | 480 | |||
M37584.1 | - | 5 | 419 | |||
X52316.1 | - | 5 | 390 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AY074806.
Panther Classification:
Gene Ontology Categories:
