siRNA Details
| Gene Symbol: Ube2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022690.2 | NP_073181.1 | 4 | 609 | ||
| GenBank mRNA | AF099093.1 | - | 4 | 576 | ||
| BC086980.1 | - | 4 | 609 | 7192141 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2g1| Gene Symbol: Ube2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022690.2 | NP_073181.1 | 4 | 609 | ||
| GenBank mRNA | AF099093.1 | - | 4 | 576 | ||
| BC086980.1 | - | 4 | 609 | 7192141 | ||