siRNA Details
| Gene Symbol: Sh3bp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022693.4 | NP_073184.3 | 4 | 2255 | ||
| XM_008767298.1 | XP_008765520.1 | 2 | 2901 | |||
| GenBank mRNA | AJ278266.2 | - | 2 | 1930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3bp4| Gene Symbol: Sh3bp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022693.4 | NP_073184.3 | 4 | 2255 | ||
| XM_008767298.1 | XP_008765520.1 | 2 | 2901 | |||
| GenBank mRNA | AJ278266.2 | - | 2 | 1930 | ||