siRNA Details
| Gene Symbol: Snd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022694.2 | NP_073185.2 | 9, 10 | 1244 | ||
| GenBank mRNA | AY697864.1 | - | 9, 10 | 1254 | ||
| BC072471.1 | - | 9, 10 | 1244 | 7100501 | ||
| U83883.1 | - | 9, 10 | 988 | |||

Snd1
Genomic Map