siRNA Details
| Gene Symbol: Taok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022702.1 | NP_073193.1 | 15, 16 | 2198 | ||
| XM_006230329.2 | XP_006230391.1 | 15, 16 | 3639 | |||
| XM_006230330.2 | XP_006230392.1 | 14, 15 | 2060 | |||
| XM_006230331.2 | XP_006230393.1 | 15, 16 | 2927 | |||
| XM_006230332.2 | XP_006230394.1 | 15, 16 | 2910 | |||
| XM_006230334.2 | XP_006230396.1 | 15, 16 | 2912 | |||
| GenBank mRNA | AB290408.1 | - | 14, 15 | 2024 | ||
| AF140556.2 | - | 15, 16 | 2198 | |||
| FQ223093.1 | - | 15, 16 | 2835 | |||

Taok2
Genomic Map