siRNA Details
| Gene Symbol: Olah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022705.1 | NP_073196.1 | 6 | 845 | ||
| XM_008771886.1 | XP_008770108.1 | 7 | 999 | |||
| XM_008771887.1 | XP_008770109.1 | 6 | 702 | |||
| GenBank mRNA | M16200.1 | - | 6 | 845 | ||
| Y00311.1 | - | 5 | 580 | |||

Olah
Genomic Map