siRNA Details
| Gene Symbol: GNA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004297.3 | NP_004288.1 | 6 | 1347 | ||
| XM_011519224.1 | XP_011517526.1 | 5 | 584 | |||
| XM_017015341.1 | XP_016870830.1 | 6 | 875 | |||
| GenBank mRNA | AF105201.1 | - | 6 | 962 | ||
| AF493903.1 | - | 6 | 824 | |||
| AK312460.1 | - | 6 | 1347 | |||
| BC027886.1 | - | 6 | 1338 | 5221889 | ||

GNA14
Genomic Map