siRNA Details
| Gene Symbol: Grin2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022797.1 | NP_073634.1 | 6 | 2193 | ||
| XM_008759316.1 | XP_008757538.1 | 5 | 1898 | |||
| XM_008759317.1 | XP_008757539.1 | 5 | 1898 | |||
| GenBank mRNA | D13213.1 | - | 5 | 1487 | ||
| L31611.1 | - | 5 | 1468 | |||
| L31612.1 | - | 6 | 2193 | |||
| U08260.1 | - | 6 | 1539 | |||

Grin2d
Genomic Map