siRNA Details
| Gene Symbol: Nucks1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022799.2 | NP_073636.1 | 6 | 674 | ||
| XM_006249797.2 | XP_006249859.1 | 6 | 743 | |||
| GenBank mRNA | AJ237669.1 | - | 5 | 499 | ||
| BC078818.1 | - | 6 | 674 | 7128528 | ||
| FQ228159.1 | - | 6 | 699 | |||

Nucks1
Genomic Map