siRNA Details
| Gene Symbol: Dpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022850.1 | NP_074041.1 | 4 | 649 | ||
| XM_006235880.2 | XP_006235942.1 | 4 | 4722 | |||
| GenBank mRNA | M76426.1 | - | 4 | 649 | ||
| M76427.1 | - | 4 | 502 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpp6| Gene Symbol: Dpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022850.1 | NP_074041.1 | 4 | 649 | ||
| XM_006235880.2 | XP_006235942.1 | 4 | 4722 | |||
| GenBank mRNA | M76426.1 | - | 4 | 649 | ||
| M76427.1 | - | 4 | 502 | |||