siRNA Details
| Gene Symbol: Tspy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022923.1 | NP_075212.1 | 3 | 830 | ||
| XM_008773672.1 | XP_008771894.1 | 3 | 837 | |||
| GenBank mRNA | AF074880.1 | - | 3 | 830 | ||
| AJ001377.1 | - | 3 | 241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspy1| Gene Symbol: Tspy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022923.1 | NP_075212.1 | 3 | 830 | ||
| XM_008773672.1 | XP_008771894.1 | 3 | 837 | |||
| GenBank mRNA | AF074880.1 | - | 3 | 830 | ||
| AJ001377.1 | - | 3 | 241 | |||