siRNA Details
| Gene Symbol: Chrna9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022930.1 | NP_075219.1 | 5 | 1211 | ||
| GenBank mRNA | AY574257.1 | - | 5 | 1124 | ||
| U12336.1 | - | 5 | 1211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna9| Gene Symbol: Chrna9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022930.1 | NP_075219.1 | 5 | 1211 | ||
| GenBank mRNA | AY574257.1 | - | 5 | 1124 | ||
| U12336.1 | - | 5 | 1211 | |||